Learn More
Sigma Aldrich Fine Chemicals Biosciences TPPR tissue culture dishes
Catalog No. 502908549
Encompass
Innovative features and outstanding cell growth properties make the TPP tissue culture dishes the ideal choice for adherent cell culture in research and industry. These optically clear high-grade polystyrene tissue culture dishes are tissue culture treated only on the base of the dish with a non-treated ring surrounding the growing surface to prevent cell attachment on the edge. A serrated ring around the bottom culture dish makes pick-up and handling of the dishes safe and easy while also minimizing the risk of dropping dishes due to lid/base disassembly. A special stacking ring with spaces for a numeric scale (12 3 6 9) set on the periphery allows for safe and stable stacking of these tissue culture dishes. The numeric scale serves to ease locating areas of examination while the spaces they create in the stacking ring reduce condensation between stacked dishes and prevent the tissue culture dishes from sticking together. Two marking areas located on the top dish wall (yellow)
Catalog No. 50-290-8549 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z707678840EA
Innovative features and outstanding cell growth properties make the TPP tissue culture dishes the ideal choice for adherent cell culture in research and industry. These optically clear high-grade polystyrene tissue culture dishes are tissue culture treated only on the base of the dish with a non-treated ring surrounding the growing surface to prevent cell attachment on the edge. A serrated ring around the bottom culture dish makes pick-up and handling of the dishes safe and easy while also minimizing the risk of dropping dishes due to lid/base disassembly. A special stacking ring with spaces for a numeric scale (12 3 6 9) set on the periphery allows for safe and stable stacking of these tissue culture dishes. The numeric scale serves to ease locating areas of examination while the spaces they create in the stacking ring reduce condensation between stacked dishes and prevent the tissue culture dishes from sticking together. Two marking areas located on the top dish wall (yellow)