Learn More
Sigma Aldrich Fine Chemicals Biosciences TPPR tissue culture plates
Catalog No. 502906016
Encompass
TPP Tissue Culture PlatesAdvantages of the TPP tissue culture platesLid with air-venting system guarantees controlled gas exchange with low evaporationSloped edge allows placement of the lid in one position onlyYellow marking area on lid and plate ensure correct lid orientation yellow on yellow - matchRidged grip area ensures a secure grasp and prevents from accidentally lifting off the lidClear alpha-numerical labelling between the wells for easy orientation during microscopy easy to see black alpha-numerics on the sideGrowth area on spherical zone onlyAbsolutely flat growth surfaceAll TPP plate sizes (6 12 24 and 96-well) stack securely togetherAir vents in plate base prevent condensation and provide consistent air-flow between stacked platesCrystal clear transparencyAdvantages of the TPP 96 well tissue culture platesManufactured according to ANSI/SBS-3d-standardsStandardized with an interleaved barcode 2/5White base
Catalog No. 50-290-6016 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z70776772EA
TPP Tissue Culture PlatesAdvantages of the TPP tissue culture platesLid with air-venting system guarantees controlled gas exchange with low evaporationSloped edge allows placement of the lid in one position onlyYellow marking area on lid and plate ensure correct lid orientation yellow on yellow - matchRidged grip area ensures a secure grasp and prevents from accidentally lifting off the lidClear alpha-numerical labelling between the wells for easy orientation during microscopy easy to see black alpha-numerics on the sideGrowth area on spherical zone onlyAbsolutely flat growth surfaceAll TPP plate sizes (6 12 24 and 96-well) stack securely togetherAir vents in plate base prevent condensation and provide consistent air-flow between stacked platesCrystal clear transparencyAdvantages of the TPP 96 well tissue culture platesManufactured according to ANSI/SBS-3d-standardsStandardized with an interleaved barcode 2/5White base