Learn More
Medchemexpress LLC HY-101294 5mg Medchemexpress, TPPU CAS:1222780-33-7 Purity:>98%
SDP
Catalog No. 501967825
Encompass_Preferred
Medchemexpress, HY-101294 5mg TPPU CAS:1222780-33-7 TTPU is a soluble epoxide hydrolase (sEH) inhibitor with IC50 values of 37 and 3.7 nM for monkey and human sEH, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7825 Supplier Medchemexpress LLC Supplier No. HY1012945MG
Medchemexpress, HY-101294 5mg TPPU CAS:1222780-33-7 TTPU is a soluble epoxide hydrolase (sEH) inhibitor with IC50 values of 37 and 3.7 nM for monkey and human sEH, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.