Learn More
Selleck Chemical LLC TPPU E2847-5MG
SDP
Catalog No. 5000040336
Encompass_Preferred
TPPU is a soluble epoxide hydrolase (sEH) inhibitor with IC50 values of 37 and 3 7 nM for monkey and human sEH respectively
Catalog No. 50-000-40336 Supplier Selleck Chemical LLC Supplier No. E28475MG
TPPU is a soluble epoxide hydrolase (sEH) inhibitor with IC50 values of 37 and 3 7 nM for monkey and human sEH respectively