Learn More
Aviva Systems Biology Corp TPR Antibody - C-terminal region (ARP61102_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61102_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TPR. Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-252