Learn More
Medchemexpress LLC TPST1 Human HEK293 5ug | 5ug
SDP
Catalog No. 5000186981
Encompass_Preferred
TPST1 (tyrosyl protein sulfotransferase 1) catalyzes the O-sulfation of tyrosine residues located within the acidic motifs of polypeptides utilizing 3 -phosphoadenylate sulfate (PAPS) as a co-substrate This enzymatic activity plays a crucial role in post-translational modification processes especially sulfation which contributes to the structural and functional diversity of proteins TPST1 Protein Human (HEK293 His) is the recombinant human-derived TPST1 expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-86981 Supplier Medchemexpress LLC Supplier No. HYP772555UG
TPST1 (tyrosyl protein sulfotransferase 1) catalyzes the O-sulfation of tyrosine residues located within the acidic motifs of polypeptides utilizing 3 -phosphoadenylate sulfate (PAPS) as a co-substrate This enzymatic activity plays a crucial role in post-translational modification processes especially sulfation which contributes to the structural and functional diversity of proteins TPST1 Protein Human (HEK293 His) is the recombinant human-derived TPST1 expressed by HEK293 with N-His labeled tag