Learn More
Medchemexpress LLC TPT-260 | 2154425-45-1 | >98% | C₃₀H₃₂FN₅O₅ | 100 MG
SDP
Catalog No. 5000354474
Encompass_Preferred
TPT-260 is a potent, selective, and orally active ATP-competitive PI3Kδ inhibitor. It potently inhibits PI3Kδ activity with a high degree of selectivity over other PI3K isoforms and kinases.
  • Potent PI3Kδ inhibitor
  • Selective inhibition over other PI3K isoforms and kinases
  • Orally active
  • ATP-competitive
Catalog No. 50-003-54474 Supplier Medchemexpress LLC Supplier No. HY144993100MG
TPT-260 is a potent, selective, and orally active ATP-competitive PI3Kδ inhibitor. It potently inhibits PI3Kδ activity with a high degree of selectivity over other PI3K isoforms and kinases.
  • Potent PI3Kδ inhibitor
  • Selective inhibition over other PI3K isoforms and kinases
  • Orally active
  • ATP-competitive