Learn More
Medchemexpress LLC TPT-260 dihydrochloride | 2076-91-7 | 98.0% | C8H14Cl2N4S3 | 10MG
SDP
Catalog No. 5000259459
Encompass_Preferred
TPT-260 Dihydrochloride is a thiourea-derived small-molecule research compound supplied as a light yellow to yellow solid with high reported purity. It is used in preclinical biochemical and pharmacological studies where controlled solubility and defined storage stability are required.
  • High purity: 98.0% ensuring reproducible results.
  • Light yellow solid with defined appearance for easy identification.
  • Well characterized solubility in common solvents (DMSO and water); ultrasonic assistance recommended.
  • Clear storage guidance for stability in solid and solution states.
  • Available in small pack sizes suitable for research-scale experiments.
Catalog No. 50-002-59459 Supplier Medchemexpress LLC Supplier No. HY13769A10MG
TPT-260 Dihydrochloride is a thiourea-derived small-molecule research compound supplied as a light yellow to yellow solid with high reported purity. It is used in preclinical biochemical and pharmacological studies where controlled solubility and defined storage stability are required.
  • High purity: 98.0% ensuring reproducible results.
  • Light yellow solid with defined appearance for easy identification.
  • Well characterized solubility in common solvents (DMSO and water); ultrasonic assistance recommended.
  • Clear storage guidance for stability in solid and solution states.
  • Available in small pack sizes suitable for research-scale experiments.