Learn More
Cayman Chemical TPT-260hydrochlorIde 10mg
SDP
Catalog No. 502973108
Encompass_Preferred
A thiophene thiourea derivative that acts as a chaperone to stabilize the retromer complex against thermal denaturation (Kd = ~5 µM); dose-dependently increases the level of retromer proteins, redirects APP from the endosome, and reduces the formation of pathogenic APP in cultured hippocampal neurons
Catalog No. 50-297-3108 Supplier Cayman Chemical Supplier No. 1607910 MG
A thiophene thiourea derivative that acts as a chaperone to stabilize the retromer complex against thermal denaturation (Kd = ~5 µM); dose-dependently increases the level of retromer proteins, redirects APP from the endosome, and reduces the formation of pathogenic APP in cultured hippocampal neurons