Learn More
Cayman Chemical TPT-260 (hydrochloride), Purity: ≥95%, CAS: 2076-91-7, 5 mg
SDP
Catalog No. 502969510
Encompass_Preferred
A thiophene thiourea derivative that acts as a chaperone to stabilize the retromer complex against thermal denaturation (Kd = ~5 µM); dose-dependently increases the level of retromer proteins, redirects APP from the endosome, and reduces the formation of pathogenic APP in cultured hippocampal neurons
Catalog No. 50-296-9510 Supplier Cayman Chemical Supplier No. 160795 MG
A thiophene thiourea derivative that acts as a chaperone to stabilize the retromer complex against thermal denaturation (Kd = ~5 µM); dose-dependently increases the level of retromer proteins, redirects APP from the endosome, and reduces the formation of pathogenic APP in cultured hippocampal neurons