Learn More
Aviva Systems Biology Corp TPTE Antibody - middle region (ARP35585_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35585_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TPTE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TPTE. Alternate names: PTEN2, CT44ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55785