Learn More
Hach Company TPTZ Iron Reagent Powder Pillows, 25mL, pk/100
Catalog No. 501995706 Shop All Hach Company Products
Encompass
For low range Iron determination by the TPTZ method. Hach Method 8112. Range: 0.012 - 1.800 mg/L Fe. Sample Size: 25 mL. Pack of 100 Powder Pillows
Catalog No. 50-199-5706 Supplier Hach Company Supplier No. 2275699
For low range Iron determination by the TPTZ method. Hach Method 8112. Range: 0.012 - 1.800 mg/L Fe. Sample Size: 25 mL. Pack of 100 Powder Pillows