Learn More
Cole-Parmer TPV Pharma Grade Tubing, 1/16" ID x 3/16" OD; 50 Ft
Supplier:  Cole-Parmer 5011982
Encompass_Preferred
Use the biocompatible TPV tubing for peristaltic pumping, filling, and fluid transfer applications. This flexible opaque tubing offers excellent pump performance while providing great chemical resistance, making it a great alternative to silicone tubing.
TPV tubing does not contain BPA, phthalates, PVC, or peroxide that can leach out into the fluid, making it perfect for pharmaceutical, laboratory, and diagnostic applications. For an added level of particle control, a liner is placed between the tubing package and the box.
Catalog No. 50-256-3443