Learn More
Medchemexpress LLC Tqs | 353483-92-8 | MFCD01080566 | 99.8% | 376.47 | C22H20N2O2S | 5 MG
SDP
Catalog No. 5000240986
Encompass_Preferred
TQS is a small-molecule positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR) intended for preclinical research. It is supplied as a high-purity solid and is characterized for solubility and storage to support both in vitro and in vivo studies.
  • Alpha7 nicotinic acetylcholine receptor positive allosteric modulator.
  • Useful in neuroinflammatory pain and neuronal signaling research.
  • High purity solid suitable for in vitro and in vivo experiments.
  • Soluble in DMSO and formulatable for in vivo dosing.
  • Stable when stored as powder at -20°C or 4°C per storage guidance.
Catalog No. 50-002-40986 Supplier Medchemexpress LLC Supplier No. HY1076825MG
TQS is a small-molecule positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR) intended for preclinical research. It is supplied as a high-purity solid and is characterized for solubility and storage to support both in vitro and in vivo studies.
  • Alpha7 nicotinic acetylcholine receptor positive allosteric modulator.
  • Useful in neuroinflammatory pain and neuronal signaling research.
  • High purity solid suitable for in vitro and in vivo experiments.
  • Soluble in DMSO and formulatable for in vivo dosing.
  • Stable when stored as powder at -20°C or 4°C per storage guidance.