Learn More
Selleck Chemical LLC TR-107 E1986-5MG
SDP
Catalog No. 5000039167
Encompass_Preferred
TR-107 is a chemical activator of the human mitochondrial protease ClpP with an EC50 of 140 nM It exhibits excellent potency and specificity in the research of triple-negative breast cancer (TNBC) cell models
Catalog No. 50-000-39167 Supplier Selleck Chemical LLC Supplier No. E19865MG
TR-107 is a chemical activator of the human mitochondrial protease ClpP with an EC50 of 140 nM It exhibits excellent potency and specificity in the research of triple-negative breast cancer (TNBC) cell models