Learn More
Medchemexpress LLC HY-15770 5mg Medchemexpress, TR-14035 CAS:232271-19-1 Purity:>98%
SDP
Catalog No. 502081795
Encompass_Preferred
Medchemexpress, HY-15770 5mg TR-14035 CAS:232271-19-1 TR-14035 is a dual α4β7/α4β1 integrin antagonist, with IC50 s of 7 nM and 87 nM for α4β7 and α4β1, respectively. TR-14035 can be used for the research of inflammation and autoimmune disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1795 Supplier Medchemexpress LLC Supplier No. HY157705MG
Medchemexpress, HY-15770 5mg TR-14035 CAS:232271-19-1 TR-14035 is a dual α4β7/α4β1 integrin antagonist, with IC50 s of 7 nM and 87 nM for α4β7 and α4β1, respectively. TR-14035 can be used for the research of inflammation and autoimmune disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.