Learn More
Foxx Life Sciences EZBiopure Titanium Round Media Bottle Assembly, MTO, 1L, PETG Bottle for Aseptic Sampling and Sterile Storage, Single Use Technology with Pureweld Tubing, CPC MPC and GL45 VersaCap with Vent and Dip Tube, 10/CS, Sterile
Catalog No. 5000077927
Encompass_Preferred
Foxx Life Sciences presents its sterile, single-use EZBiopure Titanium Round Media Bottle Assembly, available in Polyethylene Terephthalate Glycol (PETG) or Polystyrene (PS).
Designed for fluid transfer and storage, this assembly features an angled shoulder for smooth pouring and molded grips for a secure hold, even with gloves. With Foxx's patented VersaCap, these assemblies need no machining, ideal for single-use applications.
Each assembly includes EZBiopure TR Bottle, Open Cap with Adapter, Close Cap for Storage, TPE Tubing (ID 1/8” × OD ¼”), Fittings with Male & Female Connectors(CPC MPC/AQS or Plug), Diptube for Precise Draw, 0.22µm Vent Filter, Ratchet-Style Pinch Clamp. Each unit is double-bagged and Gamma Sterilized in the USA.
Features
Easy to read graduation marks with ± 5% accuracy
Made with USP Class VI components.
Gamma Compatible and meets sterile assurance level (SAL) of 10-6
Available in 125ml, 250ml, 500ml and 1L.
Catalog No. 50-000-77927 Supplier Foxx Life Sciences Supplier No. 5277451FLSX
Foxx Life Sciences presents its sterile, single-use EZBiopure Titanium Round Media Bottle Assembly, available in Polyethylene Terephthalate Glycol (PETG) or Polystyrene (PS).
Designed for fluid transfer and storage, this assembly features an angled shoulder for smooth pouring and molded grips for a secure hold, even with gloves. With Foxx's patented VersaCap, these assemblies need no machining, ideal for single-use applications.
Each assembly includes EZBiopure TR Bottle, Open Cap with Adapter, Close Cap for Storage, TPE Tubing (ID 1/8” × OD ¼”), Fittings with Male & Female Connectors(CPC MPC/AQS or Plug), Diptube for Precise Draw, 0.22µm Vent Filter, Ratchet-Style Pinch Clamp. Each unit is double-bagged and Gamma Sterilized in the USA.
Features
Easy to read graduation marks with ± 5% accuracy
Made with USP Class VI components.
Gamma Compatible and meets sterile assurance level (SAL) of 10-6
Available in 125ml, 250ml, 500ml and 1L.