Learn More
Foxx Life Sciences EZBiopure Titanium Round Media Bottle, 500mL, PETG, single use wide mouth bottle, GL45, No Cap, Non-Sterile, 24/CS
GREENER_CHOICE
Catalog No. 502906886
Encompass_Preferred
EZBiopure Titanium Round (TR) Media Bottles combine ergonomic design with pharmaceutical-grade safety. Constructed from crystal-clear USP Class VI PETG, these bottles are ideal for storing and sampling APIs, bulk intermediates, and culture buffers. Their specialized construction ensures stability for the long-term storage of pH-sensitive liquids, providing a versatile solution for laboratory and bioprocessing applications.
Features:
Angled shoulders ensure complete pouring, while a lower profile provides superior stability.
Molded finger grips allow for secure handling, even with gloved hands.
Features easy-to-read graduation marks with 5% accuracy.
Bottles can withstand low temperatures down to 70 C
Available Sizes: 125mL, 250mL, 500mL, 1000mL
Material: BPA free PETG (USP Class VI)
Gamma Compatible
Catalog No. 50-290-6886 Supplier Foxx Life Sciences Supplier No. 5071931FLS
EZBiopure Titanium Round (TR) Media Bottles combine ergonomic design with pharmaceutical-grade safety. Constructed from crystal-clear USP Class VI PETG, these bottles are ideal for storing and sampling APIs, bulk intermediates, and culture buffers. Their specialized construction ensures stability for the long-term storage of pH-sensitive liquids, providing a versatile solution for laboratory and bioprocessing applications.
Features:
Angled shoulders ensure complete pouring, while a lower profile provides superior stability.
Molded finger grips allow for secure handling, even with gloved hands.
Features easy-to-read graduation marks with 5% accuracy.
Bottles can withstand low temperatures down to 70 C
Available Sizes: 125mL, 250mL, 500mL, 1000mL
Material: BPA free PETG (USP Class VI)
Gamma Compatible