Learn More
Foxx Life Sciences EZBio Titanium Round (TR) 5L (1 Gal) Carboy, Polycarbonate (PC), Autoclavable, No Spigot, VersaCap 83B, Round Mixing carboy with Graduation Marks, Double Bagged, Non-Sterile, 1/EA
Catalog No. 502906901
Encompass_Preferred
EZBio Titanium Round (TR) Carboys are sterile fluid containers designed for high-performance bioprocessing and pharmaceutical-grade applications. The round shape ensures uniform mixing, eliminating material buildup found in square designs. These cleanroom-compatible carboys support critical fluid handling in controlled environments.
Features:
Available in 5L, 10L, and 20L sizes (Sterile and Non Sterile)
Molded from USP Class VI resins
FDA 21 CFR 177 compliant materials
Clear body for easy visual inspection
Graduation marks in liters and gallons
Leakproof VersaCap 83B compatibility
Resistant to low temperatures
Autoclavable
Manufactured in a Class 7 cleanroom
Catalog No. 50-290-6901 Supplier Foxx Life Sciences Supplier No. 518CF51FLS
EZBio Titanium Round (TR) Carboys are sterile fluid containers designed for high-performance bioprocessing and pharmaceutical-grade applications. The round shape ensures uniform mixing, eliminating material buildup found in square designs. These cleanroom-compatible carboys support critical fluid handling in controlled environments.
Features:
Available in 5L, 10L, and 20L sizes (Sterile and Non Sterile)
Molded from USP Class VI resins
FDA 21 CFR 177 compliant materials
Clear body for easy visual inspection
Graduation marks in liters and gallons
Leakproof VersaCap 83B compatibility
Resistant to low temperatures
Autoclavable
Manufactured in a Class 7 cleanroom