Learn More
Foxx Life Sciences EZBio Titanium Round (TR) 5L (1 Gal) Carboy, Polypropylene (PP), Autoclavable, No Spigot, VersaCap 83B, Round Mixing Carboy with Graduation Marks, Wide Mouth, Double Bagged, Sterile, 1/CS
Catalog No. 502906884
Encompass_Preferred
EZBio Titanium Round (TR) Carboys are sterile fluid containers designed for high-performance bioprocessing and pharmaceutical-grade applications. The round shape ensures uniform mixing, eliminating material buildup found in square designs. These cleanroom-compatible carboys support critical fluid handling in controlled environments.
Features:
Available in 5L, 10L, and 20L sizes (Sterile and Non Sterile)
Molded from USP Class VI resins
FDA 21 CFR 177 compliant materials
Graduation marks in liters and gallons
Leakproof VersaCap 83B compatibility
Autoclavable
Manufactured in a Class 7 cleanroom
Catalog No. 50-290-6884 Supplier Foxx Life Sciences Supplier No. 515CF52FLS
EZBio Titanium Round (TR) Carboys are sterile fluid containers designed for high-performance bioprocessing and pharmaceutical-grade applications. The round shape ensures uniform mixing, eliminating material buildup found in square designs. These cleanroom-compatible carboys support critical fluid handling in controlled environments.
Features:
Available in 5L, 10L, and 20L sizes (Sterile and Non Sterile)
Molded from USP Class VI resins
FDA 21 CFR 177 compliant materials
Graduation marks in liters and gallons
Leakproof VersaCap 83B compatibility
Autoclavable
Manufactured in a Class 7 cleanroom