Learn More
AURORA BIOLABS LLC TR-FRET Cereblon-4C Binding Assay Kit,384 rxn
Catalog No. NC4007636
Encompass_Preferred
E3 ubiquitin ligases are enzymes that catalyze the final step of the ubiquitination process and play essential roles in targeted protein degradation. Cereblon (CRBN) is the substrate-recognition component of the CRBN/DDB1/CUL4A/RBX1 E3 ubiquitin ligase complex and is widely used in PROTAC (Proteolysis-Targeting Chimera) drug discovery for targeted protein degradation applications. The TR-FRET Cereblon-4C Binding Assay Kit is designed to quantitatively measure the binding interaction between CRBN and its ligands. The assay includes Tag1-CRBN-4C complex, terbium-labeled anti-Tag1 antibody, and fluorescently labeled thalidomide ligand. Binding of CRBN to thalidomide generates a fluorescence resonance energy transfer (FRET) signal detected by measuring fluorescence emission at 665 nm and 620 nm. Compounds that compete with thalidomide for CRBN binding reduce the HTRF signal, enabling evaluation of CRBN-targeting molecules for drug discovery and PROTAC research applications.
Catalog No. NC4007636 Supplier AURORA BIOLABS LLC Supplier No. 272625384
E3 ubiquitin ligases are enzymes that catalyze the final step of the ubiquitination process and play essential roles in targeted protein degradation. Cereblon (CRBN) is the substrate-recognition component of the CRBN/DDB1/CUL4A/RBX1 E3 ubiquitin ligase complex and is widely used in PROTAC (Proteolysis-Targeting Chimera) drug discovery for targeted protein degradation applications. The TR-FRET Cereblon-4C Binding Assay Kit is designed to quantitatively measure the binding interaction between CRBN and its ligands. The assay includes Tag1-CRBN-4C complex, terbium-labeled anti-Tag1 antibody, and fluorescently labeled thalidomide ligand. Binding of CRBN to thalidomide generates a fluorescence resonance energy transfer (FRET) signal detected by measuring fluorescence emission at 665 nm and 620 nm. Compounds that compete with thalidomide for CRBN binding reduce the HTRF signal, enabling evaluation of CRBN-targeting molecules for drug discovery and PROTAC research applications.