Learn More
AURORA BIOLABS LLC Aurora Biolabs, TR-FRET, Cereblon-4C Binding Assay, 384 Reactions
Catalog No. NC3915361
Encompass_Preferred
The TR-FRET Cereblon-4C Binding Assay Kit is designed to measure binding affinity between Cereblon (CRBN) and its ligands. Utilizing the CRBN/DDB1/CUL4A/RBX1 complex, this assay employs TR-FRET technology with a terbium-labeled anti-Tag1 antibody and a fluorescently labeled thalidomide ligand. Quantitative measurement of the binding interaction is achieved through an HTRF signal, calculated from the ratio of emission intensities at 665 nm and 620 nm. The kit is intended for research use only.
  • Measures binding affinity between the substrate-recognition component and ligands
  • Detects inhibition of binding interactions when test compounds compete with thalidomide
  • Quantifies results via HTRF signal ratio calculation
  • Components include a protein complex, terbium-labeled antibody, and fluorophore-labeled ligand
Catalog No. NC3915361 Supplier AURORA BIOLABS LLC Supplier No. 272625
The TR-FRET Cereblon-4C Binding Assay Kit is designed to measure binding affinity between Cereblon (CRBN) and its ligands. Utilizing the CRBN/DDB1/CUL4A/RBX1 complex, this assay employs TR-FRET technology with a terbium-labeled anti-Tag1 antibody and a fluorescently labeled thalidomide ligand. Quantitative measurement of the binding interaction is achieved through an HTRF signal, calculated from the ratio of emission intensities at 665 nm and 620 nm. The kit is intended for research use only.
  • Measures binding affinity between the substrate-recognition component and ligands
  • Detects inhibition of binding interactions when test compounds compete with thalidomide
  • Quantifies results via HTRF signal ratio calculation
  • Components include a protein complex, terbium-labeled antibody, and fluorophore-labeled ligand