Learn More
AURORA BIOLABS LLC TR-FRET PARP2 Trapping Assay Kit,384 rxn
Supplier:  AURORA BIOLABS LLC 72772TAK384
Encompass_Preferred
PARP2 (Poly ADP-ribose Polymerase 2) is a DNA repair enzyme in the PARP family that plays an important role in repairing DNA single-strand breaks and maintaining genome stability. PARP2 detects DNA damage, becomes activated, and uses NAD⁺ to generate poly (ADP-ribose) chains through a process called PARylation, which recruits additional DNA repair proteins to damaged sites. PARP2 is overexpressed in several cancers and contributes to tumor cell survival. Certain PARP inhibitors not only block PARP2 catalytic activity but also trap PARP2 on damaged DNA, preventing DNA repair and leading to accumulation of toxic DNA-protein complexes. This effect is especially important in BRCA1/2-mutant cancers. The TR-FRET PARP2 Trapping Assay Kit is designed to quantitatively measure PARP2-DNA trapping activity and evaluate compounds that modulate PARP2 trapping for cancer research and drug discovery applications.
Catalog No. NC4007657