Learn More
Perkin Elmer US LLC TR+ MEINHARD Plus Glass Nebulizer for Optima/Avio and NexION 300/350
Catalog No. 502117211
Encompass_Preferred
This MEINHARD nebulizer features a self aspiration rate of 0.5 mL/min with argon flow 1 L/min at 50 PSI (3.4 bar/345 kPa). Each nebulizer comes with the FitKit+ gas and sample quick connect fittings. The connections are made with high-purity PFA, minimizing the chance of contamination during backflow events. The gas connection fits snugly over the maria on the sidearm of the nebulizer.
Catalog No. 50-211-7211 Supplier Perkin Elmer US LLC Supplier No. N8122462
This MEINHARD nebulizer features a self aspiration rate of 0.5 mL/min with argon flow 1 L/min at 50 PSI (3.4 bar/345 kPa). Each nebulizer comes with the FitKit+ gas and sample quick connect fittings. The connections are made with high-purity PFA, minimizing the chance of contamination during backflow events. The gas connection fits snugly over the maria on the sidearm of the nebulizer.