Learn More
Medchemexpress LLC Tr-PEG2-OH 1g | 105589-77-3 | 1 G
SDP
Catalog No. 5000227712
Encompass_Preferred
Tr-PEG2-OH is a PEG-based linker used to connect small molecules or payloads in PROTAC and ADC synthesis. It is a non-cleavable, two-unit polyethylene glycol spacer that increases solubility and provides flexible spacing for bioconjugation.
  • Used as a PROTAC linker and as a non-cleavable two-unit PEG ADC linker.
  • Provides spacing and improves solubility between conjugated moieties.
  • White to off-white solid with low water content (~0.31%).
  • High HPLC purity reported on COA (99.45% for batch example).
  • Available in small pack sizes suitable for research (e.g., 1 G).
Catalog No. 50-002-27712 Supplier Medchemexpress LLC Supplier No. HY1149951G
Tr-PEG2-OH is a PEG-based linker used to connect small molecules or payloads in PROTAC and ADC synthesis. It is a non-cleavable, two-unit polyethylene glycol spacer that increases solubility and provides flexible spacing for bioconjugation.
  • Used as a PROTAC linker and as a non-cleavable two-unit PEG ADC linker.
  • Provides spacing and improves solubility between conjugated moieties.
  • White to off-white solid with low water content (~0.31%).
  • High HPLC purity reported on COA (99.45% for batch example).
  • Available in small pack sizes suitable for research (e.g., 1 G).