Learn More
Medchemexpress LLC Tr-PEG2-OH 5g | 105589-77-3 | 5 G
SDP
Catalog No. 5000229553
Encompass_Preferred
Tr-PEG2-OH is a trityl-protected two-unit polyethylene glycol (PEG) linker terminating in a hydroxyl group, designed for use as a spacer in PROTAC and antibody-drug conjugate (ADC) linker chemistry. It provides solubility and flexibility while enabling controlled functionalization for conjugation and synthetic workflows.
  • Two-unit polyethylene glycol spacer for flexible linkages
  • Trityl-protected hydroxyl enables selective deprotection
  • Suitable for PROTAC and ADC linker assembly and conjugation
  • High reported purity (99.45%) from lot COA
  • Available in lab-scale quantities for synthetic use
Catalog No. 50-002-29553 Supplier Medchemexpress LLC Supplier No. HY1149955G
Tr-PEG2-OH is a trityl-protected two-unit polyethylene glycol (PEG) linker terminating in a hydroxyl group, designed for use as a spacer in PROTAC and antibody-drug conjugate (ADC) linker chemistry. It provides solubility and flexibility while enabling controlled functionalization for conjugation and synthetic workflows.
  • Two-unit polyethylene glycol spacer for flexible linkages
  • Trityl-protected hydroxyl enables selective deprotection
  • Suitable for PROTAC and ADC linker assembly and conjugation
  • High reported purity (99.45%) from lot COA
  • Available in lab-scale quantities for synthetic use