Learn More
Medchemexpress LLC Tr-PEG3-OH | 133699-09-9 | MFCD22574815 | 99.6% | 392.49 g/mol | C25H28O4 | 1 G
SDP
Catalog No. 5000202421
Encompass_Preferred
Tr-PEG3-OH is a non-cleavable three-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. It acts as a short, hydrophilic spacer to improve payload solubility and control spacing between payload and antibody or ligand, supporting efficient conjugation chemistry.
  • Non-cleavable three-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Provides a short, hydrophilic spacer to improve solubility and reduce aggregation.
  • High purity for consistent conjugation performance.
  • Compatible with standard organic synthesis and conjugation workflows.
  • Available in small research quantities for laboratory use.
Catalog No. 50-002-02421 Supplier Medchemexpress LLC Supplier No. HY1202581G
Tr-PEG3-OH is a non-cleavable three-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. It acts as a short, hydrophilic spacer to improve payload solubility and control spacing between payload and antibody or ligand, supporting efficient conjugation chemistry.
  • Non-cleavable three-unit PEG linker suitable for ADC and PROTAC synthesis.
  • Provides a short, hydrophilic spacer to improve solubility and reduce aggregation.
  • High purity for consistent conjugation performance.
  • Compatible with standard organic synthesis and conjugation workflows.
  • Available in small research quantities for laboratory use.