Learn More
OTT Hydromet Adcon - TR1 - Analog
Catalog No. NC1919656
Encompass_Preferred
On the tip of the tube you will find two highly accurate sensor elements for temperature and relative humidity, while the center of the tube is home to a signal amplifier, converting the sensors output to two standard analog signals of 0 to 2.5V DC. This sensor is virtually maintenance free and maintains its accuracy over a long period of time. A threaded cap with a wire mesh shield protects the sensor elements against dust, bugs and other pollutants. For applications requiring special protection, e.g. against aggressive dust particles like ammonia in chicken sheds we also offer optional steel sinter, PTFE sinter and PTFE membrane filter caps. Please note that we have selected the wire filter cap for its combination of two desirable effects: quick reaction to changing humidity and temperature while still providing sufficient protection against small particles.
Catalog No. NC1919656 Supplier OTT Hydromet Supplier No. 200.733.031
On the tip of the tube you will find two highly accurate sensor elements for temperature and relative humidity, while the center of the tube is home to a signal amplifier, converting the sensors output to two standard analog signals of 0 to 2.5V DC. This sensor is virtually maintenance free and maintains its accuracy over a long period of time. A threaded cap with a wire mesh shield protects the sensor elements against dust, bugs and other pollutants. For applications requiring special protection, e.g. against aggressive dust particles like ammonia in chicken sheds we also offer optional steel sinter, PTFE sinter and PTFE membrane filter caps. Please note that we have selected the wire filter cap for its combination of two desirable effects: quick reaction to changing humidity and temperature while still providing sufficient protection against small particles.