Learn More
ABclonal Technology TR1/THRB Rabbit mAb
SDP
Catalog No. 5000005294
Encompass_Preferred
The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine It is one of the several receptors for thyroid hormone and has been shown to mediate the biological activities of thyroid hormone Knockout studies in mice suggest that the different receptors while having certain extent of redundancy may mediate different functions of thyroid hormone Mutations in this gene are known to be a cause of generalized thyroid hormone resistance (GTHR) a syndrome characterized by goiter and high levels of circulating thyroid hormone (T3-T4) with normal or slightly elevated thyroid stimulating hormone (TSH) Several alternatively spliced transcript variants encoding the same protein have been observed for this gene
Catalog No. 50-000-05294 Supplier ABclonal Technology Supplier No. A22560100UL
The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine It is one of the several receptors for thyroid hormone and has been shown to mediate the biological activities of thyroid hormone Knockout studies in mice suggest that the different receptors while having certain extent of redundancy may mediate different functions of thyroid hormone Mutations in this gene are known to be a cause of generalized thyroid hormone resistance (GTHR) a syndrome characterized by goiter and high levels of circulating thyroid hormone (T3-T4) with normal or slightly elevated thyroid stimulating hormone (TSH) Several alternatively spliced transcript variants encoding the same protein have been observed for this gene