Learn More
MOUSER ELECTRONICS TR30RAM150-12E03-GY-BK-LEVEL-V
Supplier:  MOUSER ELECTRONICS 418TR30RAM150V12
Encompass
Wall Mount AC Adapters Switching Adapter, Level V, Wallmount, Medical, 30 Watt, 90-264VAC Input, 15VDC Output, 5.5x2.5x12mm, Straight, DC Cable 1800mm, Grey Case
Catalog No. NC2044621