Learn More
STEMCELL Technologies Anti-Human TRA-1-60 Antibody, Clone TRA-1-60R, PE, Size: 100 Tests
SDP
Supplier:  STEMCELL Technologies 60064PE
Encompass
Clone TRA-1-60R is a mouse monoclonal antibody that reacts with TRA-1-60 (podocalyxin) a 200 kDa pluripotent stem cell-specific protein expressed on the surface of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells embryonal carcinoma cells and embryonic germ cells as well as rhesus monkey ES cell lines. Verified for use with cell separation flow cytometry immunocytochemistry immunofluorescence microscopy and western blotting assays use this antibody in applications requiring the detection of TRA-1-60. This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR-E8 (Catalog 05940) mTeSR1 (Catalog 85850) and TeSR2 (Catalog 05860) and for purity assessments of cells isolated with EasySep Human ES/iPS Cell TRA-1-60 Positive Selection Kit (Catalog 18166 partial blocking may be observed) and EasySep hESC/hiPSC SSEA-4 Positive Selection Kit (Cata
Catalog No. 50-239-0275