Learn More
Aviva Systems Biology Corp TRABD Antibody - middle region (ARP42648_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42648_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TRABD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRABD. Alternate names: LP6054, MGC110928, PP2447ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58291