Learn More
Medchemexpress LLC HY-50936 5mg , Trabectedin CAS:114899-77-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY509365MG
Encompass_Preferred
HY-50936 5mg Trabectedin CAS: 114899-77-3 Trabectedin (Ecteinascidin 743; ET-743) is a tetrahydroisoquinoline alkaloid with potent antitumor activity. Trabectedin binds to the minor groove of DNA, blocks transcription of stress-induced proteins, induces DNA backbone cleavage and cancer cells apoptosis, and increases the generation of ROS in MCF-7 and MDA-MB-453 cells. Trabectedin has the potential for soft tissue sarcoma and ovarian cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0262