Learn More
Medchemexpress LLC Trabectedin-d3 (Ecteinascidin 743-d3) | 114899-77-3 | 99.1% | C39H40D3N3O11S | 1 MG
SDP
Catalog No. 5000358368
Encompass_Preferred
Trabectedin D3 (Ecteinascidin 743 D3) is deuterium labeled Trabectedin, a tetrahydroisoquinoline alkaloid with potent antitumor activity. It binds to the minor groove of DNA, blocking transcription of stress-induced proteins, inducing DNA backbone cleavage, and cancer cell apoptosis. It also increases the generation of reactive oxygen species (ROS) in MCF-7 and MDA-MB-453 cells. This product has potential for soft tissue sarcoma and ovarian cancer treatment and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-58368 Supplier Medchemexpress LLC Supplier No. HY50936S1MG
Trabectedin D3 (Ecteinascidin 743 D3) is deuterium labeled Trabectedin, a tetrahydroisoquinoline alkaloid with potent antitumor activity. It binds to the minor groove of DNA, blocking transcription of stress-induced proteins, inducing DNA backbone cleavage, and cancer cell apoptosis. It also increases the generation of reactive oxygen species (ROS) in MCF-7 and MDA-MB-453 cells. This product has potential for soft tissue sarcoma and ovarian cancer treatment and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.