Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tracazolate hydrochloride | 1135210-68-2 | 99.2% | 5 MG
SDP

Catalog No. 5000412848
Encompass_Preferred

Tracazolate (ICI 136753) hydrochloride is a potent GABAA receptor modulator with selectivity for β3, exhibiting anxiolytic and anticonvulsant activity. Its potency is determined by the nature of the third subunit (γ1-3, δ, ε) within the receptor complex. This compound is for research use only and not sold to patients.

  • Potentiates α1β1γ2s (EC50=13.2 μM) and α1β3γ2 (EC50=1.5 μM).
  • Inhibits α1β1ε (EC50=4.0 μM), α1β3ε (EC50=1.2 μM), α1β3 (EC50=2.7 μM), and α6β3γ (EC50=1.1 μM).
  • Interacts with γ-aminobutyric acid GABAA receptors, adenosine receptors, and phosphodiesterases.
  • Increases maximum current amplitude and enhances sensitivity of α1β2δ-containing GABAA receptors in oocytes at 10 μM.

Catalog No. 50-004-12848 Supplier Medchemexpress LLC Supplier No. HYB1803A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tracazolate (ICI 136753) hydrochloride is a potent GABAA receptor modulator with selectivity for β3, exhibiting anxiolytic and anticonvulsant activity. Its potency is determined by the nature of the third subunit (γ1-3, δ, ε) within the receptor complex. This compound is for research use only and not sold to patients.

  • Potentiates α1β1γ2s (EC50=13.2 μM) and α1β3γ2 (EC50=1.5 μM).
  • Inhibits α1β1ε (EC50=4.0 μM), α1β3ε (EC50=1.2 μM), α1β3 (EC50=2.7 μM), and α6β3γ (EC50=1.1 μM).
  • Interacts with γ-aminobutyric acid GABAA receptors, adenosine receptors, and phosphodiesterases.
  • Increases maximum current amplitude and enhances sensitivity of α1β2δ-containing GABAA receptors in oocytes at 10 μM.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.