Learn More
Medchemexpress LLC Tracazolate hydrochloride | 1135210-68-2 | MFCD03791317 | 99.2% | 340.85 g/mol | C16H25ClN4O2 | 10 MG
SDP
Catalog No. 5000298997
Encompass_Preferred
Tracazolate hydrochloride is the hydrochloride salt of tracazolate (ICI 136753), a potent positive allosteric modulator of GABAA receptors with reported subtype selectivity. It is supplied as a research reagent for in vitro and in vivo studies of GABAA receptor pharmacology, anxiolytic effects, and anticonvulsant activity.
  • Potent GABAA receptor positive allosteric modulator.
  • Reported selectivity for the β3 subunit.
  • High purity (99.2%).
  • Off-white to light yellow solid appearance.
  • Recommended storage: -20°C, sealed, protect from moisture.
  • Available in small research pack sizes such as 10 MG.
Catalog No. 50-002-98997 Supplier Medchemexpress LLC Supplier No. HYB1803A10MG
Tracazolate hydrochloride is the hydrochloride salt of tracazolate (ICI 136753), a potent positive allosteric modulator of GABAA receptors with reported subtype selectivity. It is supplied as a research reagent for in vitro and in vivo studies of GABAA receptor pharmacology, anxiolytic effects, and anticonvulsant activity.
  • Potent GABAA receptor positive allosteric modulator.
  • Reported selectivity for the β3 subunit.
  • High purity (99.2%).
  • Off-white to light yellow solid appearance.
  • Recommended storage: -20°C, sealed, protect from moisture.
  • Available in small research pack sizes such as 10 MG.