Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tracazolate hydrochloride, Tocris Bioscience™

Catalog No. 155810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-581-0 10 mg
15-585-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-581-0 Supplier R&D Systems Supplier No. 1558/10
This item is not returnable. View return policy

Allosteric modulator of GABAA receptors

GABAA receptor modulator. Can potentiate or inhibit recombinant GABAA function depending on subunit combination. Enhances native GABAA receptor function and is anxiolytic following systemic administration in vivo.

Chemical Identifiers

CAS 1135210-68-2
Molecular Formula C16H25ClN4O2
Molecular Weight (g/mol) 340.852
InChI Key TUIXDPUEMQXVHO-UHFFFAOYSA-N
Synonym tracazolate hydrochloride, dsstox_cid_25208, dsstox_rid_80751, dsstox_gsid_45208, cas-nocas_45208, 4-butylamino-1-ethyl-6-methyl-1h-pyrazolo 3,4-b pyridine-5-carboxylic acid ethyl ester
PubChem CID 24871267
IUPAC Name ethyl 4-(butylamino)-1-ethyl-6-methylpyrazolo[3,4-b]pyridine-5-carboxylate;hydrochloride
SMILES CCCCNC1=C2C=NN(C2=NC(=C1C(=O)OCC)C)CC.Cl

Specifications

Quantity 10 mg
Formula Weight 340.85
Percent Purity >99%
Chemical Name or Material Tracazolate hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.