Learn More
Description
- Selenium aids in the detoxification of free radicals as a cofactor for GSH synthetase, while iron, copper, and zinc may be bound by serum protein
- The combination of Trace Elements A contains the trace elements of MCDB 104 and of MCDB 301
Specifications
Specifications
| Concentration | 1000x |
| Form | Solution |
| Product Type | Trace Element |
| Sterility | Sterile-filtered |
| Sterilization Method | Sterile-filtered |
| Quantity | 1 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.