Learn More
Description
- Vinyl backing cards are intended for use with trace evidence collection tape.
- Helps prevent contamination and loss of collected evidence.
- Allows convenient labeling on the reverse side.
Specifications
Specifications
| Product Type | Backing Card |
| For Use With (Application) | Protection of Collected Evidence from Contamination or Loss |
| Color | Black |
| Quantity | 25/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
