Learn More
Cole-Parmer Trace Metal Analysis Pipette Tips, Non-Sterile, 101 to 1000 µL; 10 Racks of 100 Tips
Catalog No. NC9535377 Shop All Cole-Parmer Products
Encompass_Preferred
These universal tips range in volume from 1 to 1000 µL and fit a variety of pipettors to ensurelaboratory standardization. The 200 µL tips are beveled to increase accuracy and reproducibility. A reference mark at 10 µL lets you check pipettor calibration and visually estimate sample size. Tips are packaged in racks for easy access.
Catalog No. NC9535377 Supplier Cole-Parmer Supplier No. 2571300
These universal tips range in volume from 1 to 1000 µL and fit a variety of pipettors to ensurelaboratory standardization. The 200 µL tips are beveled to increase accuracy and reproducibility. A reference mark at 10 µL lets you check pipettor calibration and visually estimate sample size. Tips are packaged in racks for easy access.