Learn More
Cole-Parmer Cole-Parmer 113-96RGN Trace Metal Analysis Tips, 1 to 200 µL
Catalog No. NC1585455 Shop All Cole-Parmer Products
Encompass_Preferred
These universal tips range in volume from 1 to 1000 µL and fit a variety of pipettors to ensurelaboratory standardization. The 200 µL tips are beveled to increase accuracy and reproducibility. A reference mark at 10 µL lets you check pipettor calibration and visually estimate sample size. Tips are packaged in racks for easy access.
Catalog No. NC1585455 Supplier Cole-Parmer Supplier No. 2571302
These universal tips range in volume from 1 to 1000 µL and fit a variety of pipettors to ensurelaboratory standardization. The 200 µL tips are beveled to increase accuracy and reproducibility. A reference mark at 10 µL lets you check pipettor calibration and visually estimate sample size. Tips are packaged in racks for easy access.