Learn More
ERA WATERS CORP TRACE METALS
SDP
Catalog No. NC9778225
Encompass
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with AA, ICP-OES or ICP-MS and selected colorimetric methods.
Catalog No. NC9778225 Supplier ERA WATERS CORP Supplier No. 586
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with AA, ICP-OES or ICP-MS and selected colorimetric methods.