Learn More
ATCC Trace Mineral Supplement for Bacteriological Culture Media, Package of 5 (10 mL/vial) (ATCC MD-TMS™)
Supplier:  ATCC MDTMS
Encompass
Trace Mineral Supplement (ATCC MD-TMS™) is a sterile additive for bacteriological culture media, especially for extremophile cultivation. Based on Wolfe’s formulation, it includes EDTA, MgSO4, MnSO4, FeSO4, ZnSO4, CuSO4, and other essential trace elements. Supplied as 5 vials (10 mL each).
Catalog No. 50-238-3325