Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Trace Minerals for an Ultra-Clean Environment

Catalog No. MP29602645 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP29602645 500 g
ICN96026401 1 kg
MP29602641 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP29602645 Supplier MP Biomedicals™ Supplier No. 02960264.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trace Minerals is used in ultra clean environments where trace elements usually present, are absent.

  • 7-trace elements which are usually present in a open environment
  • Research animal diet supplement
  • Powdered
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
For Use With (Application) Research animal diet supplements
Quantity 500 g
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.