Learn More
Cole-Parmer TRACEABLE CAL ALARM THERMOTIME
Supplier:  Cole-Parmer 9876725
Encompass_Preferred
Traceable Calibrated Alarm Thermometer/Timer with Stainless Steel Probe. Key Features: Includes NIST-Traceable certificate from A2LA accredited 1750.01 Calibration laboratory.Unique adjustable display and large digits allow for easy viewing.Two distinctive alarms signal time and temperature. Features a stainless steel probe with silicone coated cable. Cable withstands up to 260 degree C.
Catalog No. NC3525569