Learn More
Perkin Elmer US LLC Traceable Calibration Kit - DMA 8000
Supplier:  Perkin Elmer US LLC N5330319
Encompass_Preferred
The DMA 8000 comes with a complete set of measuring fixtures to allow tension, bending (3 types), shear, and compression experiments. PerkinElmer has made the components of these fixtures available for individual purchase to keep them performing optimally.
Catalog No. 50-210-4345