Learn More
Research Products International Corp Traceable Cip-It Timer
SDP
Catalog No. 50999176
Encompass
Timer features count down; alarm and stopwatch functions. Timing range is 1 second to 100 hour, with 1 second resolution. Stopwatch mode is 1 second to 99 hours with repeat.
Catalog No. 50-999-176 Supplier Research Products International Corp Supplier No. 810055
Timer features count down; alarm and stopwatch functions. Timing range is 1 second to 100 hour, with 1 second resolution. Stopwatch mode is 1 second to 99 hours with repeat.