Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Digital-Bottle™ Refrigerator/Freezer Thermometers
SCI_ED

Catalog No. S02274 Shop All Fisherbrand Products
Change view
Click to view available options
Bottle Fill:
Glass Beads
Liquid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottle Fill
S02274 Liquid
S02276 Glass Beads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02274
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Chemical resistant ABS Plastic holder stand, double back tape, Velcro™ fastener and replaceable Silver Oxide battery, Stainless Steel probe sealed in nontoxic Glycol filled bottle

Accurately monitor temperature in freezers, refrigerators, incubators and environmental chambers with this temperature-buffered sensor.

Provide accurate readings everywhere.

  • Sensor is insulated from rapid changes when refrigerator door is opened
  • Stainless-steel sensor/probe assures long product life
  • Entire unit is waterproof
  • Records high/low readings: Easy-to-read digital display shows high, low and current temperatures
  • Supplied with unique holder: Chemical-resistant ABS plastic holder stands on a shelf or holder affixes to any wall

Digital-Bottle eliminates mercury contamination:

  • Solid-state probe eliminates the difficulty encountered with mercury thermometer contamination
  • Probe is sealed in a miniature bottle filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA
  • Also available with glass-bead fill

Specifications:

  • Range: -20° to +50°C (-4° to +122°F)
  • Resolution: 0.1°
  • Accuracy: ±1°C (Ultra™ model: ±0.4°C at tested points)

Order Info

Overall: 1.87 x 1 x 4.62 in. (4.8 x 2.5 x 11.7cm). Holder: 2.25 x 2.25 x 1.12 in. (5.7 x 5.7 x 2.9cm). Weight: 3.67 oz. (0.1kg).

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Bottle Fill Liquid
Certifications/Compliance ISO 17025, A2LA, NIST
Depth (English) 1 in.
Depth (Metric) 2.5 cm
Height (English) 4.8 in.
Height (Metric) 12.19 cm
Includes Chemical resistant ABS Plastic holder stand, double back tape, Velcro™ fastener and replaceable Silver Oxide battery, Stainless Steel probe sealed in nontoxic Glycol filled bottle
Length (Metric) 21 cm
Material Plastic
Model Standard Model
Probe Type Sealed Probe
Resolution 0.1°C
Weight (English) 3.67 oz.
Weight (Metric) 100 g
Width (English) 1.87 in.
Product Type Digital Thermometer
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) -4°F
Min. Temperature (Metric) -20°C
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Calibrated Yes
For Use With (Equipment) Freezer, refrigerator, incubator and environmental chamber
Accuracy ±1°C
Display Digital
Width (Metric) 4.8 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.