Learn More
Research Products International Corp Traceable Laboratory Giant Digit Timer, with Certificate of Calibration and Battery
SDP
Catalog No. 501978488
Encompass
Convenient laboratory timer with giant size digits over 1" high that are visible from a distance of up to fifty feet. Convenient three button operation starts and stops countdown timing cycle on and counts up during alarm. Timing range is 99 minutes and 59 seconds with 0.01% accuracy. Also includes memory reset feature.
Supplied with certificate of calibration and battery.
Catalog No. 50-197-8488 Supplier Research Products International Corp Supplier No. 815011
Convenient laboratory timer with giant size digits over 1" high that are visible from a distance of up to fifty feet. Convenient three button operation starts and stops countdown timing cycle on and counts up during alarm. Timing range is 99 minutes and 59 seconds with 0.01% accuracy. Also includes memory reset feature.
Supplied with certificate of calibration and battery.