Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Traceable™ Hi-Accuracy 0.01° Refrigerator Thermometer
SCI_ED

Catalog No. S98173 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
2 x Bottle Probes
2 x Probes
Bottle Probe
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S98173 Bottle Probe
S98174 2 x Bottle Probes
S98175 2 x Probes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S98173
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For monitoring temperatures in refrigerator and freezer simultaneously or two refrigerator locations. Traceable™ Hi-Accuracy 0.01° Refrigerator Thermometer permits user one-button self-calibration which reads to 0.01° while simultaneously monitoring temperatures in refrigerator and freezer.

Triple display simultaneously shows minimum/maximum and current temperatures where minimum/maximum monitors readings overnight, on weekends or for any time period, and displays the exact time and date.

  • Temperature-buffered bottle sensor
  • Two visual (LEDs) and two audio alerts when temperature rises above or falls below high/low set points
  • Programmable alarms in 0.10° increments
  • Miniature bottle sealing: 1 x 2.5 in. (2.5 x 6.3 cm) filled with nontoxic glycol
  • High-impact, chemical-resistant ABS plastic case
  • Stand, Velcro™, magnetic strips, and wall mount allow placement of thermometer anywhere
  • Used in water baths, heating blocks and incubators

Specifications

  • Range: -58°F to +158°F and -50°C to +70°C
  • Resolution: 0.01°
  • Accuracy: ±0.3°C
  • Weight: 5 oz. (141 g)
  • Includes 2 AA batteries for continuous, one-year, always-on monitoring

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Dimensions (L x W x H) 4.5 x 2.75 x 0.75 in. (11.4 x 6.9 x 1.9 cm)
For Use With (Application) Monitoring temperature in refrigerator and freezer simultaneously or two refrigerator locations
Height (English) 0.75 in.
Height (Metric) 1.9 cm
Includes Bottle Probe
Length (English) Cable 120 in.
Length (English) Probe 0.7 in.
Length (Metric) 114 mm
Length (Metric) Cable 304.8 cm
Length (Metric) Probe 1.9 cm
Material ABS Plastic
Resolution 0.01°C
Weight (English) 5 oz.
Weight (Metric) 140 g
Width (English) 2.75 in.
Product Type Digital Thermometer
Length (English) 4.5 in.
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
Battery Type AA
Accuracy ±0.3°C
Display LED
Width (Metric) 6.9 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.